Assay Details
Target Gene Details
Entrez Gene ID: | 79587 |
Gene Name: | cysteinyl-tRNA synthetase 2, mitochondrial (putative) |
Gene Aliases: |
COXPD27, cysRS |
Location: |
Chr.13:110641410-110713527 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CARS2 | NM_024537.3 | NP_078813.1 | ||
XM_006719953.2 | XP_006720016.1 | |||
XM_011521116.1 | XP_011519418.1 | |||
XM_017020741.1 | XP_016876230.1 | |||
XM_017020742.1 | XP_016876231.1 | |||
AB062436.1 | BAB93499.1 | |||
AK022180.1 | BAB13978.1 | |||
AK091992.1 | ||||
AK092947.1 | ||||
AK301907.1 | ||||
BC007220.1 | AAH07220.1 | |||
BX648882.1 | CAH10378.1 | |||
BX649191.1 | CAE46210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv563125 | Chr.13:110627655 - 110657270 on Build GRCh38 | Loss |
![]() ![]() |
nsv1044234 | Chr.13:110490706 - 110718233 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1042232 | Chr.13:110629776 - 110696128 on Build GRCh38 | Gain |
![]() ![]() |
nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)