Assay Details
Target Gene Details
Entrez Gene ID: | 90627 |
Gene Name: | StAR related lipid transfer domain containing 13 |
Gene Aliases: |
ARHGAP37, DLC2, GT650, LINC00464 |
Location: |
Chr.13:33103135-33676835 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD13 | NM_001243466.1 | NP_001230395.1 | ||
NM_001243474.1 | NP_001230403.1 | |||
NM_001243476.2 | NP_001230405.1 | |||
NM_052851.2 | NP_443083.1 | |||
NM_178006.3 | NP_821074.1 | |||
NM_178007.2 | NP_821075.1 | |||
XM_011535299.2 | XP_011533601.1 | |||
XM_017020834.1 | XP_016876323.1 | |||
XM_017020835.1 | XP_016876324.1 | |||
AK091804.1 | ||||
AK094704.1 | ||||
AK095114.1 | ||||
AK312888.1 | ||||
AL049801.1 | CAB42562.1 | |||
AY082589.1 | AAL91648.1 | |||
AY082590.1 | AAL91649.1 | |||
AY082591.1 | AAL91650.1 | |||
AY366448.1 | AAQ72791.1 | |||
BC046563.1 | AAH46563.1 | |||
BX647695.1 | CAI46026.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |