Assay Details
Target Gene Details
Entrez Gene ID: | 688 |
Gene Name: | Kruppel like factor 5 |
Gene Aliases: |
BTEB2, CKLF, IKLF |
Location: |
Chr.13:73054976-73077542 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF5 | NM_001286818.1 | NP_001273747.1 | ||
NM_001730.4 | NP_001721.2 | |||
AB030824.1 | BAA96461.1 | |||
AF132818.1 | AAF18307.1 | |||
AF287272.1 | AAF88068.1 | |||
AK296900.1 | ||||
AK302280.1 | ||||
AK304284.1 | ||||
BC007695.1 | AAH07695.1 | |||
BC042131.1 | AAH42131.1 | |||
BT006641.1 | AAP35287.1 | |||
D14520.1 | BAA03393.1 | |||
EF208215.1 | ||||
HF546203.1 | ||||
HF546204.1 | ||||
HQ628639.1 | ||||
HQ628640.1 | ||||
HQ628641.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |