Assay Details
Target Gene Details
Entrez Gene ID: | 65244 |
Gene Name: | spermatogenesis associated serine rich 2 |
Gene Aliases: |
Nbla00526, P59SCR, SCR59, SPATA10 |
Location: |
Chr.12:49366905-49527441 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATS2 | NM_001293285.1 | NP_001280214.1 | ||
NM_001293286.1 | NP_001280215.1 | |||
NM_023071.3 | NP_075559.2 | |||
NR_120663.1 | ||||
XM_011538690.2 | XP_011536992.1 | |||
XM_011538691.2 | XP_011536993.1 | |||
XM_017019840.1 | XP_016875329.1 | |||
AK023202.1 | BAB14460.1 | |||
AK095582.1 | ||||
AK292334.1 | ||||
AL833614.1 | ||||
BC048299.1 | AAH48299.1 | |||
BX648916.1 | ||||
CN421379.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3580270 | Chr.12:49409096 - 49418729 on Build GRCh38 | Loss | SPATS2 |
nsv832405 | Chr.12:49326496 - 49517079 on Build GRCh38 | Gain | LOC100335030 SPATS2 TROAP DNAJC22 C1QL4 |
esv3629458 | Chr.12:49408725 - 49415138 on Build GRCh38 | Loss | SPATS2 |
dgv1497n100 | Chr.12:49284746 - 49438835 on Build GRCh38 | Gain | PRPH LOC101927267 LOC100335030 SPATS2 TROAP DNAJC22 C1QL4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |