Assay Details
Target Gene Details
Entrez Gene ID: | 217 |
Gene Name: | aldehyde dehydrogenase 2 family (mitochondrial) |
Gene Aliases: |
ALDH-E2, ALDHI, ALDM |
Location: |
Chr.12:111766887-111809985 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH2 | NM_000690.3 | NP_000681.2 | ||
NM_001204889.1 | NP_001191818.1 | |||
AK223373.1 | BAD97093.1 | |||
AK301375.1 | ||||
AK314856.1 | ||||
AY621070.1 | AAT41621.1 | |||
BC002967.1 | AAH02967.1 | |||
BC071839.1 | AAH71839.1 | |||
CR456991.1 | CAG33272.1 | |||
DC353712.1 | ||||
X05409.1 | CAA28990.1 | |||
Y00109.1 | CAA68290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2760710 | Chr.12:111735404 - 111876910 on Build GRCh38 | Gain | MIR6761 ALDH2 MAPKAPK5 MAPKAPK5-AS1 ACAD10 |
dgv514e212 | Chr.12:111735392 - 111884067 on Build GRCh38 | Gain | MIR6761 ALDH2 MAPKAPK5 MAPKAPK5-AS1 ACAD10 |
dgv304e214 | Chr.12:111742954 - 111877939 on Build GRCh38 | Gain | MIR6761 ALDH2 MAPKAPK5 MAPKAPK5-AS1 ACAD10 |
nsv470317 | Chr.12:111761276 - 111868361 on Build GRCh38 | Gain | MIR6761 ALDH2 MAPKAPK5 MAPKAPK5-AS1 |
dgv1556n100 | Chr.12:111732560 - 111876898 on Build GRCh38 | Gain | MIR6761 ALDH2 MAPKAPK5 MAPKAPK5-AS1 ACAD10 |
dgv2874n54 | Chr.12:111743274 - 111884680 on Build GRCh38 | Gain | MIR6761 ALDH2 MAPKAPK5 MAPKAPK5-AS1 ACAD10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |