Assay Details
Target Gene Details
Entrez Gene ID: | 171017 |
Gene Name: | zinc finger protein 384 |
Gene Aliases: |
CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1 |
Location: |
Chr.12:6666477-6689572 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761725 | Chr.12:6682182 - 6754772 on Build GRCh38 | Gain | PIANP MLF2 COPS7A ZNF384 |
nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss | PIANP SCARNA11 MLF2 COPS7A ING4 CHD4 ZNF384 ACRBP LPAR5 |
nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss | PIANP MLF2 CDCA3 PTMS ING4 ZNF384 P3H3 LAG3 GNB3 USP5 COPS7A CD4 GPR162 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |