Assay Details
Target Gene Details
Entrez Gene ID: | 83468 |
Gene Name: | glycosyltransferase 8 domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.12:103988983-104064177 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLT8D2 | NM_001316967.1 | NP_001303896.1 | ||
NM_031302.4 | NP_112592.1 | |||
XM_011538793.2 | XP_011537095.1 | |||
XM_017019999.1 | XP_016875488.1 | |||
AJ271711.1 | CAC19667.1 | |||
AK027310.1 | BAB55033.1 | |||
AK056056.1 | BAB71085.1 | |||
AY358763.1 | AAQ89123.1 | |||
BC022343.1 | AAH22343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv560039 | Chr.12:103783890 - 104061562 on Build GRCh38 | Gain | TTC41P TDG C12orf73 GLT8D2 NT5DC3 MIR3652 HSP90B1 |
esv3581849 | Chr.12:104006503 - 104028420 on Build GRCh38 | Gain | GLT8D2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |