Assay Details
Target Gene Details
Entrez Gene ID: | 5893 |
Gene Name: | RAD52 homolog, DNA repair protein |
Gene Aliases: |
- |
Location: |
Chr.12:911028-991195 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD52 | NM_001297419.1 | NP_001284348.1 | ||
NM_001297421.1 | NP_001284350.1 | |||
NM_001297422.1 | NP_001284351.1 | |||
NM_134424.3 | NP_602296.2 | |||
NR_123713.1 | ||||
XM_005253720.4 | XP_005253777.1 | |||
XM_005253721.2 | XP_005253778.1 | |||
XM_011520990.2 | XP_011519292.1 | |||
XM_011520991.2 | XP_011519293.1 | |||
XM_011520993.2 | XP_011519295.1 | |||
XM_017019769.1 | XP_016875258.1 | |||
XM_017019770.1 | XP_016875259.1 | |||
XM_017019771.1 | XP_016875260.1 | |||
AF125948.1 | AAD24575.1 | |||
AF125949.1 | AAD24576.1 | |||
AK290047.1 | ||||
AK292160.1 | ||||
AK294709.1 | ||||
AK299283.1 | ||||
AK312026.1 | ||||
AL558884.3 | ||||
BC104015.1 | AAI04016.1 | |||
BC104017.1 | AAI04018.1 | |||
BC114954.1 | AAI14955.1 | |||
HY045122.1 | ||||
L33262.1 | AAB05203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain | RAD52 WNK1 LOC105369595 NINJ2 LOC100049716 B4GALNT3 ERC1 |
nsv1042333 | Chr.12:798809 - 972343 on Build GRCh38 | Gain | RAD52 WNK1 |
nsv556942 | Chr.12:938365 - 1010792 on Build GRCh38 | Gain | RAD52 ERC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |