Assay Details
Target Gene Details
Entrez Gene ID: | 65987 |
Gene Name: | potassium channel tetramerization domain containing 14 |
Gene Aliases: |
- |
Location: |
Chr.11:78015715-78046191 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCTD14 | NM_001282406.1 | NP_001269335.1 | ||
AK313892.1 |
Target Gene Details
Entrez Gene ID: | 100532726 |
Gene Name: | NDUFC2-KCTD14 readthrough |
Gene Aliases: |
- |
Location: |
Chr.11:78015715-78080219 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFC2-KCTD14 | NM_001203260.1 | NP_001190189.1 | ||
NM_001203261.1 | NP_001190190.1 | |||
NM_001203262.1 | NP_001190191.1 | |||
BG723290.1 | ||||
FY211285.1 | ||||
FY212410.1 | ||||
FY212967.1 | ||||
FY213449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv516638 | Chr.11:78013628 - 78052743 on Build GRCh38 | Loss |
![]() ![]() |
nsv1050891 | Chr.11:77705600 - 78076956 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)