Assay Details
Target Gene Details
Entrez Gene ID: | 56834 |
Gene Name: | G protein-coupled receptor 137 |
Gene Aliases: |
C11orf4, GPR137A, TM7SF1L1 |
Location: |
Chr.11:64270337-64289500 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR137 | NM_001170726.1 | NP_001164197.1 | ||
NM_001170881.1 | NP_001164352.1 | |||
NM_001177358.1 | NP_001170829.1 | |||
XM_005274101.2 | XP_005274158.1 | |||
XM_005274102.2 | XP_005274159.1 | |||
XM_005274104.2 | XP_005274161.1 | |||
XM_011545168.2 | XP_011543470.1 | |||
XM_011545169.1 | XP_011543471.1 | |||
XM_011545170.2 | XP_011543472.1 | |||
XM_011545171.2 | XP_011543473.1 | |||
XM_011545172.2 | XP_011543474.1 | |||
XM_017018014.1 | XP_016873503.1 | |||
XM_017018015.1 | XP_016873504.1 | |||
XM_017018016.1 | XP_016873505.1 | |||
AK300209.1 | ||||
AK302256.1 | ||||
BC035002.1 | AAH35002.1 | |||
BI830247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | BAD PPP1R14B FKBP2 NAA40 CCDC88B TEX40 LOC105369340 VEGFB NUDT22 GPR137 MACROD1 KCNK4-TEX40 PLCB3 DNAJC4 FERMT3 STIP1 TRMT112 ESRRA FLRT1 TRPT1 COX8A KCNK4 OTUB1 PRDX5 MIR7155 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | BAD PPP1R14B FKBP2 TEX40 LOC105369340 VEGFB NUDT22 GPR137 KCNK4-TEX40 TRPT1 PLCB3 DNAJC4 KCNK4 FERMT3 STIP1 TRMT112 PRDX5 ESRRA |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | BAD PPP1R14B FKBP2 CCDC88B TEX40 LOC105369340 VEGFB NUDT22 GPR137 MIR1237 KCNK4-TEX40 TRPT1 PLCB3 DNAJC4 KCNK4 FERMT3 RPS6KA4 TRMT112 PRDX5 ESRRA MIR7155 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | BAD PPP1R14B FLRT1 FKBP2 TEX40 LOC105369340 VEGFB NUDT22 GPR137 MACROD1 KCNK4-TEX40 TRPT1 PLCB3 DNAJC4 KCNK4 FERMT3 STIP1 TRMT112 PRDX5 ESRRA |
nsv357 | Chr.11:64284335 - 64308789 on Build GRCh38 | Insertion | BAD KCNK4-TEX40 KCNK4 TEX40 ESRRA GPR137 |