Assay Details
Target Gene Details
Entrez Gene ID: | 1408 |
Gene Name: | cryptochrome circadian clock 2 |
Gene Aliases: |
HCRY2, PHLL2 |
Location: |
Chr.11:45847118-45883248 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRY2 | NM_001127457.2 | NP_001120929.1 | ||
NM_021117.3 | NP_066940.2 | |||
AB014558.1 | BAA31633.1 | |||
AK291260.1 | ||||
AK294904.1 | ||||
AK302865.1 | ||||
BC035161.1 | AAH35161.1 | |||
BC041814.1 | AAH41814.1 | |||
DC330290.1 | ||||
EU219622.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |