Assay Details
Target Gene Details
Entrez Gene ID: | 11098 |
Gene Name: | protease, serine 23 |
Gene Aliases: |
SIG13, SPUVE, ZSIG13 |
Location: |
Chr.11:86791059-86952910 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRSS23 | NM_001293180.1 | NP_001280109.1 | ||
DA396057.1 | ||||
DA895063.1 | ||||
DN917578.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3627099 | Chr.11:86660723 - 87045565 on Build GRCh38 | Gain | PRSS23 TMEM135 OR7E13P OR7E2P FZD4 ME3 LOC100506368 |
nsv428263 | Chr.11:86646948 - 86797438 on Build GRCh38 | Loss | PRSS23 ME3 |
nsv521505 | Chr.11:86727073 - 86804030 on Build GRCh38 | Gain | PRSS23 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |