Assay Details
Target Gene Details
Entrez Gene ID: | 28960 |
Gene Name: | decapping enzyme, scavenger |
Gene Aliases: |
ARS, DCS1, HINT-5, HINT5, HSL1, HSPC015 |
Location: |
Chr.11:126303301-126345754 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCPS | NM_014026.4 | NP_054745.1 | ||
XM_011542778.2 | XP_011541080.1 | |||
XM_017017612.1 | XP_016873101.1 | |||
AF077201.1 | AAD26996.1 | |||
AF532613.1 | AAM90310.1 | |||
AK026554.1 | ||||
AK223089.1 | BAD96809.1 | |||
AK307403.1 | ||||
AY040771.1 | AAK91765.1 | |||
AY077684.1 | AAL77216.1 | |||
BC014532.1 | AAH14532.1 | |||
BE382749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss | ST3GAL4-AS1 SRPRA TIRAP FOXRED1 ST3GAL4 HP11014 DCPS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |