Assay Details
Target Gene Details
Entrez Gene ID: | 116985 |
Gene Name: | ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1 |
Gene Aliases: |
CENTD2 |
Location: |
Chr.11:72685069-72752389 on Build GRCh38 |
Assay Gene Location: | Within Intron 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARAP1 | NM_001040118.2 | NP_001035207.1 | ||
NM_001135190.1 | NP_001128662.1 | |||
NM_015242.4 | NP_056057.2 | |||
AB018325.2 | ||||
AF411983.1 | AAL04167.1 | |||
AJ621557.1 | CAF21317.1 | |||
AK160383.1 | BAD18726.1 | |||
AY049732.1 | AAL12169.1 | |||
AY553630.1 | ||||
BC008315.1 | AAH08315.1 | |||
BC056401.1 | AAH56401.1 | |||
BC140792.1 | ||||
BC144562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523601 | Chr.11:72570840 - 72719890 on Build GRCh38 | Loss | MIR139 LINC01537 ARAP1 PDE2A |
nsv832207 | Chr.11:72561706 - 72758077 on Build GRCh38 | Loss | MIR139 LINC01537 ARAP1 PDE2A STARD10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |