Assay Details
Target Gene Details
Entrez Gene ID: | 2915 |
Gene Name: | glutamate metabotropic receptor 5 |
Gene Aliases: |
GPRC1E, MGLUR5, PPP1R86, mGlu5 |
Location: |
Chr.11:88504576-89065971 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRM5 | NM_000842.4 | NP_000833.1 | ||
NM_001143831.2 | NP_001137303.1 | |||
XM_006718828.3 | XP_006718891.1 | |||
XM_011542792.1 | XP_011541094.1 | |||
XM_017017627.1 | XP_016873116.1 | |||
AK291362.1 | ||||
AY608336.1 | AAT37960.1 | |||
D28538.1 | BAA05891.1 | |||
D28539.1 | BAA05892.1 | |||
FJ588505.1 | ||||
FJ588506.1 | ||||
FJ588507.1 | ||||
FJ668670.1 | ||||
S64316.1 | AAD13954.1 |
Target Gene Details
Entrez Gene ID: | 100873989 |
Gene Name: | GRM5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:88504576-88524054 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRM5-AS1 | NR_049724.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035387 | Chr.11:88442010 - 88772705 on Build GRCh38 | Gain | GRM5-AS1 GRM5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |