Assay Details
Target Gene Details
Entrez Gene ID: | 747 |
Gene Name: | diacylglycerol lipase alpha |
Gene Aliases: |
C11orf11, DAGL(ALPHA), DAGLALPHA, NSDDR |
Location: |
Chr.11:61680184-61747002 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAGLA | NM_006133.2 | NP_006124.1 | ||
XM_017018238.1 | XP_016873727.1 | |||
XM_017018240.1 | XP_016873729.1 | |||
AB014559.2 | ||||
BC150176.1 | ||||
BC150195.1 | ||||
BC152453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832179 | Chr.11:61605799 - 61757528 on Build GRCh38 | Loss | RPLP0P2 DKFZP434K028 DAGLA LOC101927495 MYRF |
nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss | DAGLA FEN1 MIR1908 MIR611 TMEM258 MIR6746 DKFZP434K028 FADS2 FADS1 FADS3 MYRF RAB3IL1 LOC100129473 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |