Assay Details
Target Gene Details
Entrez Gene ID: | 2286 |
Gene Name: | FK506 binding protein 2 |
Gene Aliases: |
FKBP-13, PPIase |
Location: |
Chr.11:64240941-64244135 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP2 | NM_001135208.1 | NP_001128680.1 | ||
NM_004470.3 | NP_004461.2 | |||
NM_057092.2 | NP_476433.1 | |||
XM_005273848.3 | XP_005273905.1 | |||
BC003384.1 | AAH03384.1 | |||
BC091475.1 | AAH91475.1 | |||
BU682774.1 | ||||
M65128.1 | AAA58473.1 | |||
M75099.1 | AAA36563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | NUDT22 STIP1 PLCB3 VEGFB BAD MACROD1 DNAJC4 TRPT1 LOC105369340 FKBP2 GPR137 PPP1R14B FERMT3 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | NUDT22 PLCB3 COX8A PRDX5 VEGFB KCNK4-TEX40 BAD DNAJC4 MIR7155 TEX40 FKBP2 KCNK4 TRMT112 PPP1R14B STIP1 OTUB1 NAA40 FLRT1 MACROD1 TRPT1 LOC105369340 CCDC88B ESRRA GPR137 FERMT3 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | NUDT22 STIP1 PLCB3 PRDX5 VEGFB KCNK4-TEX40 BAD DNAJC4 TRPT1 LOC105369340 TEX40 FKBP2 ESRRA GPR137 KCNK4 TRMT112 PPP1R14B FERMT3 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | NUDT22 STIP1 PLCB3 VEGFB FKBP2 DNAJC4 PPP1R14B FERMT3 TRPT1 LOC105369340 |
esv24861 | Chr.11:64233793 - 64250642 on Build GRCh38 | Gain | VEGFB FKBP2 DNAJC4 PPP1R14B LOC105369340 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | NUDT22 PLCB3 PRDX5 VEGFB KCNK4-TEX40 BAD DNAJC4 MIR7155 TRPT1 LOC105369340 CCDC88B RPS6KA4 TEX40 FKBP2 ESRRA GPR137 KCNK4 MIR1237 TRMT112 PPP1R14B FERMT3 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | NUDT22 STIP1 PLCB3 VEGFB BAD MACROD1 DNAJC4 TRPT1 LOC105369340 FKBP2 GPR137 PPP1R14B FERMT3 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | NUDT22 STIP1 PLCB3 PRDX5 VEGFB KCNK4-TEX40 BAD FLRT1 MACROD1 DNAJC4 TRPT1 LOC105369340 TEX40 FKBP2 ESRRA GPR137 KCNK4 TRMT112 PPP1R14B FERMT3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |