Assay Details
Target Gene Details
Entrez Gene ID: | 11309 |
Gene Name: | solute carrier organic anion transporter family member 2B1 |
Gene Aliases: |
OATP-B, OATP2B1, OATPB, SLC21A9 |
Location: |
Chr.11:75150987-75206400 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLCO2B1 | NM_001145211.2 | NP_001138683.1 | ||
NM_001145212.2 | NP_001138684.1 | |||
NM_007256.4 | NP_009187.1 | |||
XM_017017157.1 | XP_016872646.1 | |||
AB020687.1 | ||||
AB026256.1 | BAA78638.1 | |||
AF205073.1 | AAG42205.1 | |||
AK290234.1 | ||||
AK294503.1 | ||||
AK300134.1 | ||||
AK302492.1 | ||||
AK304783.1 | ||||
AL117465.1 | CAB55940.1 | |||
BC041095.1 | AAH41095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054122 | Chr.11:75161747 - 75184196 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)