Assay Details
Target Gene Details
Entrez Gene ID: | 2900 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 4 |
Gene Aliases: |
EAA1, GRIK, GluK4, KA1 |
Location: |
Chr.11:120511746-120988805 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIK4 | NM_001282470.2 | NP_001269399.1 | ||
NM_001282473.2 | NP_001269402.1 | |||
NM_014619.4 | NP_055434.2 | |||
XM_011542784.1 | XP_011541086.1 | |||
XM_011542786.2 | XP_011541088.1 | |||
XM_011542787.2 | XP_011541089.1 | |||
XM_017017621.1 | XP_016873110.1 | |||
XM_017017622.1 | XP_016873111.1 | |||
AK292726.1 | ||||
AK314284.1 | ||||
BC146652.1 | ||||
BC150173.1 | ||||
S67803.1 | AAB29311.1 |
Target Gene Details
Entrez Gene ID: | 101929227 |
Gene Name: | uncharacterized LOC101929227 |
Gene Aliases: |
- |
Location: |
Chr.11:120867933-120894797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929227 | NR_132790.1 | |||
CR936773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1053607 | Chr.11:120670086 - 120931297 on Build GRCh38 | Gain | LOC101929227 GRIK4 LOC105369532 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |