Assay Details
Target Gene Details
Entrez Gene ID: | 5795 |
Gene Name: | protein tyrosine phosphatase, receptor type J |
Gene Aliases: |
CD148, DEP1, HPTPeta, R-PTP-ETA, SCC1 |
Location: |
Chr.11:47980504-48170842 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPRJ | NM_001098503.1 | NP_001091973.1 | ||
NM_002843.3 | NP_002834.3 | |||
XM_017018083.1 | XP_016873572.1 | |||
XM_017018085.1 | XP_016873574.1 | |||
AL359057.1 | CAB94390.1 | |||
BC063417.1 | AAH63417.1 | |||
D37781.1 | BAA07035.1 | |||
DR007037.1 | ||||
U10886.1 | AAB36687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528641 | Chr.11:47982817 - 48021501 on Build GRCh38 | Loss | PTPRJ |
nsv1053145 | Chr.11:47946350 - 48065398 on Build GRCh38 | Gain | PTPRJ |
dgv1111n100 | Chr.11:47653918 - 48644472 on Build GRCh38 | Gain | OR4X2 OR4S1 FNBP4 OR4C3 PTPRJ OR4X1 OR4B1 NUP160 MIR3161 AGBL2 OR4A47 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |