Assay Details
Target Gene Details
Entrez Gene ID: | 51105 |
Gene Name: | PHD finger protein 20-like 1 |
Gene Aliases: |
CGI-72, TDRD20B, URLC1 |
Location: |
Chr.8:132775358-132848807 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831463 | Chr.8:132723206 - 132905755 on Build GRCh38 | Gain | TG TMEM71 PHF20L1 |
nsv1016348 | Chr.8:132769270 - 132803626 on Build GRCh38 | Loss | PHF20L1 |
dgv7317n100 | Chr.8:132769940 - 132833582 on Build GRCh38 | Loss | PHF20L1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |