Assay Details
Target Gene Details
Entrez Gene ID: | 101926926 |
Gene Name: | RDH10 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:73310906-73356461 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RDH10-AS1 | NR_125388.1 | |||
AK128216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion | LOC392232 RPL7 MSC STAU2 TRPA1 RDH10 MSC-AS1 STAU2-AS1 LOC101926908 KCNB2 RDH10-AS1 SBSPON TERF1 C8orf89 |