Assay Details
Target Gene Details
Entrez Gene ID: | 1191 |
Gene Name: | clusterin |
Gene Aliases: |
AAG4, APO-J, APOJ, CLI, CLU1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2 |
Location: |
Chr.8:27596917-27616717 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLU | NM_001831.3 | NP_001822.3 | ||
NR_038335.1 | ||||
NR_045494.1 | ||||
XM_006716284.2 | XP_006716347.1 | |||
AK093399.1 | ||||
AK301324.1 | ||||
AK313870.1 | ||||
AY513288.1 | AAT08041.1 | |||
BC010514.1 | AAH10514.1 | |||
BC019588.2 | AAH19588.1 | |||
BX459023.2 | ||||
BX648414.1 | CAI45990.1 | |||
EB385800.1 | ||||
J02908.1 | AAA51765.1 | |||
M25915.1 | AAA35692.1 | |||
M64722.1 | AAB06508.1 | |||
M74816.1 | AAA60321.1 | |||
X14723.1 | CAA32847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3572654 | Chr.8:27600526 - 27612426 on Build GRCh38 | Loss | MIR6843 CLU |
nsv831275 | Chr.8:27470773 - 27624088 on Build GRCh38 | Loss | MIR6843 EPHX2 CLU CHRNA2 |
esv3572652 | Chr.8:27589383 - 27606101 on Build GRCh38 | Loss | EPHX2 CLU |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |