Assay Details
Target Gene Details
Entrez Gene ID: | 6674 |
Gene Name: | sperm associated antigen 1 |
Gene Aliases: |
CILD28, CT140, HEL-S-268, HSD-3.8, SP75, TPIS |
Location: |
Chr.8:100157869-100241904 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPAG1 | NM_003114.4 | NP_003105.2 | ||
NM_172218.2 | NP_757367.1 | |||
XM_011517240.2 | XP_011515542.1 | |||
XM_011517241.2 | XP_011515543.1 | |||
XM_011517242.2 | XP_011515544.1 | |||
XM_011517243.2 | XP_011515545.1 | |||
XM_011517245.2 | XP_011515547.1 | |||
XM_017013754.1 | XP_016869243.1 | |||
XM_017013755.1 | XP_016869244.1 | |||
AF311312.1 | AAG23967.1 | |||
AK057482.1 | ||||
BC055091.1 | AAH55091.1 | |||
DB028984.1 | ||||
GQ472210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv611814 | Chr.8:100097796 - 100177024 on Build GRCh38 | Loss | RGS22 FBXO43 SPAG1 POLR2K |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |