Assay Details
Target Gene Details
Entrez Gene ID: | 2167 |
Gene Name: | fatty acid binding protein 4 |
Gene Aliases: |
A-FABP, AFABP, ALBP, HEL-S-104, aP2 |
Location: |
Chr.8:81478497-81483238 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FABP4 | NM_001442.2 | NP_001433.1 | ||
BC003672.1 | AAH03672.1 | |||
BQ880795.1 | ||||
BT006809.1 | AAP35455.1 | |||
CD000452.1 | ||||
CR456903.1 | CAG33184.1 | |||
FJ224319.1 | ||||
J02874.1 | AAA51689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv611607 | Chr.8:81405621 - 81768032 on Build GRCh38 | Gain | FABP12 FABP4 SLC10A5 PMP2 IMPA1 CHMP4C ZFAND1 FABP9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |