Assay Details
Target Gene Details
Entrez Gene ID: | 2137 |
Gene Name: | exostosin like glycosyltransferase 3 |
Gene Aliases: |
BOTV, EXTL1L, EXTR1, REGR, RPR |
Location: |
Chr.8:28701473-28753690 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXTL3 | NM_001440.3 | NP_001431.1 | ||
NR_073468.1 | ||||
NR_073469.1 | ||||
XM_011544440.2 | XP_011542742.1 | |||
AB007042.1 | BAA24080.1 | |||
AB011091.1 | ||||
AF001690.1 | AAC39598.1 | |||
AF083551.1 | AAD42041.1 | |||
BI911162.1 | ||||
DA222815.1 | ||||
DA757362.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)