Assay Details
Target Gene Details
Entrez Gene ID: | 84750 |
Gene Name: | fucosyltransferase 10 |
Gene Aliases: |
FUCTX |
Location: |
Chr.8:33308102-33473492 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FUT10 | XM_011544676.2 | XP_011542978.1 | ||
XM_011544677.2 | XP_011542979.1 | |||
XM_011544678.2 | XP_011542980.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576539 | Chr.8:32789248 - 33495217 on Build GRCh38 | Gain | LOC105379362 MAK16 FUT10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |