Assay Details
Target Gene Details
Entrez Gene ID: | 64798 |
Gene Name: | DEP domain containing MTOR-interacting protein |
Gene Aliases: |
DEP.6, DEPDC6 |
Location: |
Chr.8:119873655-120056202 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DEPTOR | NM_001283012.1 | NP_001269941.1 | ||
NM_022783.3 | NP_073620.2 | |||
AK022490.1 | BAB14054.1 | |||
AK023916.1 | BAB14723.1 | |||
AK225168.1 | ||||
AK225675.1 | ||||
AK297822.1 | ||||
AK315175.1 | ||||
AL136678.1 | CAB66613.1 | |||
BC012040.1 | AAH12040.1 | |||
BC024746.1 | AAH24746.1 | |||
DA622978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv612111 | Chr.8:119624610 - 119982154 on Build GRCh38 | Gain | DSCC1 TAF2 DEPTOR ENPP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |