Assay Details
Target Gene Details
Entrez Gene ID: | 25960 |
Gene Name: | adhesion G protein-coupled receptor A2 |
Gene Aliases: |
GPR124, TEM5 |
Location: |
Chr.8:37796883-37843986 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRA2 | NM_032777.9 | NP_116166.9 | ||
XM_005273471.3 | XP_005273528.1 | |||
XM_011544481.2 | XP_011542783.1 | |||
XM_011544482.2 | XP_011542784.1 | |||
XM_011544483.2 | XP_011542785.1 | |||
AB040964.2 | BAA96055.2 | |||
AF378755.1 | AAL11992.1 | |||
AK027296.1 | BAB55022.1 | |||
AK074099.2 | ||||
AY090637.1 | AAM09084.1 | |||
AY181242.1 | AAO27354.1 | |||
BC146774.1 | ||||
KU257672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509261 | Chr.8:37767151 - 37881434 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
nsv528524 | Chr.8:37789148 - 37839356 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)