Assay Details
Target Gene Details
Entrez Gene ID: | 105379393 |
Gene Name: | uncharacterized LOC105379393 |
Gene Aliases: |
- |
Location: |
Chr.8:42051750-42104987 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105379393 | NR_136589.1 | |||
HY030269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3617000 | Chr.8:42060242 - 42072166 on Build GRCh38 | Loss | LOC105379393 |
esv3616999 | Chr.8:42059429 - 42085578 on Build GRCh38 | Loss | LOC105379393 |