Assay Details
Target Gene Details
Entrez Gene ID: | 23414 |
Gene Name: | zinc finger protein, FOG family member 2 |
Gene Aliases: |
DIH3, FOG2, SRXY9, ZC2HC11B, ZNF89B, hFOG-2 |
Location: |
Chr.8:105318859-105804539 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZFPM2 | NM_012082.3 | NP_036214.2 | ||
XM_011516948.2 | XP_011515250.1 | |||
AF119334.1 | AAD49558.1 | |||
AK296997.1 | ||||
BC109222.1 | AAI09223.1 | |||
BC109223.1 | AAI09224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521187 | Chr.8:105311508 - 105352623 on Build GRCh38 | Loss |
![]() |
nsv611837 | Chr.8:105298349 - 105359952 on Build GRCh38 | Loss |
![]() |
nsv1034972 | Chr.8:105294788 - 105357360 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)