Assay Details
Target Gene Details
Entrez Gene ID: | 55284 |
Gene Name: | ubiquitin conjugating enzyme E2 W (putative) |
Gene Aliases: |
UBC-16, UBC16 |
Location: |
Chr.8:73780097-73878910 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2W | NM_001271015.1 | NP_001257944.1 | ||
BC010900.1 | AAH10900.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |