Assay Details
Target Gene Details
Entrez Gene ID: | 25901 |
Gene Name: | coiled-coil domain containing 28A |
Gene Aliases: |
C6orf80, CCRL1AP |
Location: |
Chr.6:138773517-138793319 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC28A | NM_015439.2 | NP_056254.1 | ||
AJ420568.1 | ||||
AK300672.1 | ||||
AL050197.1 | CAB43315.1 | |||
AY167571.1 | AAN87344.1 | |||
BC004464.1 | AAH04464.1 | |||
BC013019.2 | ||||
BC080566.1 | AAH80566.1 | |||
BC108717.1 | AAI08718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5500 | Chr.6:138787783 - 138820753 on Build GRCh38 | Insertion | CCDC28A ECT2L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |