Assay Details
Target Gene Details
Entrez Gene ID: | 60481 |
Gene Name: | ELOVL fatty acid elongase 5 |
Gene Aliases: |
HELO1, SCA38, dJ483K16.1 |
Location: |
Chr.6:53267398-53349179 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ELOVL5 | NM_001242828.1 | NP_001229757.1 | ||
NM_001242830.1 | NP_001229759.1 | |||
NM_001301856.1 | NP_001288785.1 | |||
NM_021814.4 | NP_068586.1 | |||
AB209798.1 | BAD93035.1 | |||
AF231981.1 | AAF70631.1 | |||
AF338241.1 | AAM00193.1 | |||
AK074889.1 | BAC11270.1 | |||
AK125098.1 | ||||
AK299674.1 | ||||
AK302948.1 | ||||
AL136939.1 | CAB66873.1 | |||
BC067123.1 | AAH67123.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1033994 | Chr.6:53225739 - 53458464 on Build GRCh38 | Gain | MIR5685 ELOVL5 RPS16P5 GCM1 |
esv2763940 | Chr.6:53228517 - 53622284 on Build GRCh38 | Gain+Loss | MIR5685 LOC101927136 ELOVL5 GCLC RPS16P5 GCM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |