Assay Details
Target Gene Details
Entrez Gene ID: | 1839 |
Gene Name: | heparin binding EGF like growth factor |
Gene Aliases: |
DTR, DTS, DTSF, HEGFL |
Location: |
Chr.5:140332843-140346603 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HBEGF | NM_001945.2 | NP_001936.1 | ||
AK222688.1 | BAD96408.1 | |||
AK307310.1 | ||||
AK313202.1 | ||||
BC033097.1 | AAH33097.1 | |||
M60278.1 | AAA35956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509090 | Chr.5:140311306 - 140421024 on Build GRCh38 | Insertion | SLC4A9 HBEGF ANKHD1-EIF4EBP3 ANKHD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |