Assay Details
Target Gene Details
Entrez Gene ID: | 91368 |
Gene Name: | CDKN2A interacting protein N-terminal like |
Gene Aliases: |
- |
Location: |
Chr.5:134402065-134414625 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDKN2AIPNL | XM_011543709.2 | XP_011542011.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)