Assay Details
Target Gene Details
Entrez Gene ID: | 1794 |
Gene Name: | dedicator of cytokinesis 2 |
Gene Aliases: |
IMD40 |
Location: |
Chr.5:169637247-170083382 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOCK2 | NM_004946.2 | NP_004937.1 | ||
XM_005265830.3 | XP_005265887.1 | |||
XM_011534448.2 | XP_011532750.1 | |||
XM_011534449.2 | XP_011532751.1 | |||
XM_011534450.2 | XP_011532752.1 | |||
XM_011534451.2 | XP_011532753.1 | |||
XM_017009189.1 | XP_016864678.1 | |||
XM_017009190.1 | XP_016864679.1 | |||
BC104900.1 | AAI04901.1 | |||
BC113457.1 | ||||
D86964.1 | ||||
DB145764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5124 | Chr.5:169631419 - 169676697 on Build GRCh38 | Deletion | DOCK2 |
nsv1031257 | Chr.5:169272339 - 169832306 on Build GRCh38 | Gain | SPDL1 DOCK2 SLIT3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |