Assay Details
Target Gene Details
Entrez Gene ID: | 56477 |
Gene Name: | C-C motif chemokine ligand 28 |
Gene Aliases: |
CCK1, MEC, SCYA28 |
Location: |
Chr.5:43356972-43412391 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL28 | NM_001301873.1 | NP_001288802.1 | ||
NM_001301874.1 | NP_001288803.1 | |||
NM_001301875.1 | NP_001288804.1 | |||
NM_148672.3 | NP_683513.1 | |||
AF110384.1 | AAG43193.1 | |||
AF220210.1 | AAF87205.1 | |||
AF266504.1 | AAG16691.1 | |||
AK307307.1 | ||||
AK312120.1 | ||||
BC062668.1 | AAH62668.1 | |||
BC069532.1 | AAH69532.1 | |||
BX486632.1 | ||||
CD671712.1 | ||||
HM067700.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3604897 | Chr.5:43343819 - 43405322 on Build GRCh38 | Gain | CCL28 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |