Assay Details
Target Gene Details
Entrez Gene ID: | 337876 |
Gene Name: | chondroitin sulfate synthase 3 |
Gene Aliases: |
CHSY2, CSS3 |
Location: |
Chr.5:129903415-130186637 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHSY3 | NM_175856.4 | NP_787052.3 | ||
XM_005271982.3 | XP_005272039.1 | |||
XM_005271983.3 | XP_005272040.1 | |||
XM_011543363.2 | XP_011541665.1 | |||
XM_011543365.2 | XP_011541667.1 | |||
XM_017009435.1 | XP_016864924.1 | |||
AB086062.1 | BAC98832.1 | |||
AB175496.1 | ||||
AJ504664.2 | CAD43233.1 | |||
AJ578034.2 | CAE17326.1 | |||
AY726604.1 | ||||
BC137325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830488 | Chr.5:129843019 - 130014740 on Build GRCh38 | Gain | CHSY3 |
esv3890736 | Chr.5:129823851 - 130151212 on Build GRCh38 | Gain | CHSY3 |
esv2763498 | Chr.5:129423618 - 130021576 on Build GRCh38 | Gain | KIAA1024L ADAMTS19-AS1 CHSY3 ADAMTS19 |
nsv599704 | Chr.5:129170896 - 130297608 on Build GRCh38 | Gain | KIAA1024L ADAMTS19-AS1 CHSY3 MIR4460 ADAMTS19 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |