Assay Details
Target Gene Details
Entrez Gene ID: | 2255 |
Gene Name: | fibroblast growth factor 10 |
Gene Aliases: |
- |
Location: |
Chr.5:44301655-44389780 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGF10 | NM_004465.1 | NP_004456.1 | ||
XM_005248264.3 | XP_005248321.1 | |||
AB002097.1 | BAA22331.1 | |||
AF411527.1 | AAL05875.1 | |||
AF508782.1 | AAM46926.1 | |||
AK315314.1 | ||||
BC069561.1 | AAH69561.1 | |||
BC105021.1 | AAI05022.1 | |||
BC105023.1 | AAI05024.1 | |||
CR541665.1 | CAG46466.1 | |||
CR541688.1 | CAG46489.1 | |||
GQ351295.1 | ||||
U67918.1 | AAB61991.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4816 | Chr.5:44307243 - 44351910 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)