Assay Details
Target Gene Details
Entrez Gene ID: | 9315 |
Gene Name: | neuronal regeneration related protein |
Gene Aliases: |
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17 |
Location: |
Chr.5:111729303-111976931 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv599367 | Chr.5:111588583 - 112267016 on Build GRCh38 | Gain | LOC101927023 EPB41L4A-AS1 NREP NREP-AS1 STARD4-AS1 EPB41L4A SNORA13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |