Assay Details
Target Gene Details
Entrez Gene ID: | 23398 |
Gene Name: | peptidylprolyl isomerase domain and WD repeat containing 1 |
Gene Aliases: |
- |
Location: |
Chr.5:65563236-65587549 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPWD1 | NM_001278926.1 | NP_001265855.1 | ||
NM_001278927.1 | NP_001265856.1 | |||
NM_001278929.1 | NP_001265858.1 | |||
NM_015342.3 | NP_056157.1 | |||
XM_005248474.3 | XP_005248531.1 | |||
XM_006714583.3 | XP_006714646.1 | |||
XM_011543295.1 | XP_011541597.1 | |||
XM_017009286.1 | XP_016864775.1 | |||
AK025679.1 | ||||
AK298175.1 | ||||
AK300022.1 | ||||
AK301654.1 | ||||
BC015385.1 | AAH15385.1 | |||
D38552.1 | BAA07555.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)