Assay Details
Target Gene Details
Entrez Gene ID: | 64902 |
Gene Name: | alanine--glyoxylate aminotransferase 2 |
Gene Aliases: |
AGT2, DAIBAT |
Location: |
Chr.5:34998101-35048188 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGXT2 | NM_001306173.1 | NP_001293102.1 | ||
NM_031900.3 | NP_114106.1 | |||
XM_005248337.2 | XP_005248394.1 | |||
XM_005248338.2 | XP_005248395.1 | |||
XM_017009748.1 | XP_016865237.1 | |||
AB193309.1 | BAD66662.1 | |||
AJ292204.1 | CAC24841.1 | |||
AK055732.1 | ||||
AK055757.1 | ||||
AK223128.1 | BAD96848.1 | |||
AK223144.1 | ||||
AK223375.1 | BAD97095.1 | |||
AK314691.1 | ||||
BC144268.1 | ||||
BC150603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv597810 | Chr.5:34948427 - 35060217 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1028968 | Chr.5:34945017 - 35060217 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3604691 | Chr.5:34934667 - 35064045 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)