Assay Details
Target Gene Details
Entrez Gene ID: | 1004 |
Gene Name: | cadherin 6 |
Gene Aliases: |
CAD6, KCAD |
Location: |
Chr.5:31193655-31329146 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH6 | NM_004932.3 | NP_004923.1 | ||
XM_011513921.2 | XP_011512223.1 | |||
XM_017008911.1 | XP_016864400.1 | |||
AK291290.1 | ||||
AU130185.1 | ||||
BC000019.2 | AAH00019.1 | |||
DC353791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5619n100 | Chr.5:31199804 - 31471735 on Build GRCh38 | Gain | CDH6 DROSHA |
esv3604572 | Chr.5:31134943 - 31452028 on Build GRCh38 | Gain | CDH6 DROSHA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |