Assay Details
Target Gene Details
Entrez Gene ID: | 133584 |
Gene Name: | EGF like, fibronectin type III and laminin G domains |
Gene Aliases: |
AGRINL, AGRNL, PIKA |
Location: |
Chr.5:38258409-38465480 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGFLAM | NM_001205301.1 | NP_001192230.1 | ||
NM_152403.3 | NP_689616.2 | |||
AK096474.1 | BAC04800.1 | |||
AK291602.1 | ||||
BC063822.1 | AAH63822.1 | |||
BX647551.1 | CAH56137.1 |
Target Gene Details
Entrez Gene ID: | 100852408 |
Gene Name: | EGFLAM antisense RNA 4 |
Gene Aliases: |
- |
Location: |
Chr.5:38282269-38290986 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGFLAM-AS4 | NR_046219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3604766 | Chr.5:38221565 - 38446015 on Build GRCh38 | Gain | EGFLAM EGFLAM-AS4 EGFLAM-AS2 |
esv3604761 | Chr.5:38056637 - 38805862 on Build GRCh38 | Gain | LIFR-AS1 MIR3650 LINC01265 EGFLAM LIFR LOC101929745 OSMR-AS1 EGFLAM-AS4 EGFLAM-AS2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |