Assay Details
Target Gene Details
Entrez Gene ID: | 401190 |
Gene Name: | regulator of G-protein signaling 7 binding protein |
Gene Aliases: |
R7BP |
Location: |
Chr.5:64504565-64614661 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS7BP | NM_001029875.2 | NP_001025046.1 | ||
XM_005248502.3 | XP_005248559.1 | |||
AK294808.1 | ||||
AK296443.1 | ||||
BC140707.1 | ||||
BX640900.1 | CAE45947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4861 | Chr.5:64484259 - 64529368 on Build GRCh38 | Deletion | RGS7BP |
nsv830324 | Chr.5:64457623 - 64686227 on Build GRCh38 | Loss | RGS7BP |
nsv830323 | Chr.5:64375894 - 64568227 on Build GRCh38 | Gain | RGS7BP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |