Assay Details
Target Gene Details
Entrez Gene ID: | 206358 |
Gene Name: | solute carrier family 36 member 1 |
Gene Aliases: |
Dct1, LYAAT1, PAT1, TRAMD3 |
Location: |
Chr.5:151411344-151556318 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC36A1 | XM_011537591.1 | XP_011535893.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2730921 | Chr.5:151410030 - 151413192 on Build GRCh38 | Deletion |
![]() |
nsv600005 | Chr.5:151409490 - 151413016 on Build GRCh38 | Loss |
![]() |
dgv10127n54 | Chr.5:151411732 - 151414457 on Build GRCh38 | Loss |
![]() |
dgv10126n54 | Chr.5:151411622 - 151413016 on Build GRCh38 | Loss |
![]() |
dgv981n67 | Chr.5:151409445 - 151413174 on Build GRCh38 | Loss |
![]() |
esv2670257 | Chr.5:151409884 - 151414331 on Build GRCh38 | Deletion |
![]() |
nsv819196 | Chr.5:151409463 - 151413859 on Build GRCh38 | Loss |
![]() |
dgv10125n54 | Chr.5:151411076 - 151412762 on Build GRCh38 | Loss |
![]() |
nsv1137534 | Chr.5:151410439 - 151413039 on Build GRCh38 | Deletion |
![]() |
dgv10124n54 | Chr.5:151410812 - 151414457 on Build GRCh38 | Loss |
![]() |
dgv982n67 | Chr.5:151410300 - 151413174 on Build GRCh38 | Loss |
![]() |
esv3607177 | Chr.5:151410222 - 151413066 on Build GRCh38 | Loss |
![]() |
esv23145 | Chr.5:151410242 - 151413122 on Build GRCh38 | Loss |
![]() |
dgv1049e199 | Chr.5:151410172 - 151413398 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)