Assay Details
Target Gene Details
Entrez Gene ID: | 8507 |
Gene Name: | ectodermal-neural cortex 1 |
Gene Aliases: |
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10 |
Location: |
Chr.5:74627406-74641424 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENC1 | NM_001256574.1 | NP_001243503.1 | ||
NM_001256575.1 | NP_001243504.1 | |||
NM_001256576.1 | NP_001243505.1 | |||
NM_003633.3 | NP_003624.1 | |||
NR_046318.1 | ||||
XM_011543696.2 | XP_011541998.1 | |||
XM_011543697.2 | XP_011541999.1 | |||
AF005381.1 | AAC64498.1 | |||
AF010314.1 | AAC39532.1 | |||
AF059611.1 | AAC26109.1 | |||
AK096578.1 | ||||
AK295128.1 | ||||
AK299624.1 | ||||
AK312608.1 | ||||
BC000418.2 | AAH00418.1 | |||
DA032687.1 | ||||
DA498140.1 | ||||
DA785554.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |