Assay Details
Target Gene Details
Entrez Gene ID: | 55568 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 10 |
Gene Aliases: |
GALNACT10, PPGALNACT10, PPGANTASE10 |
Location: |
Chr.5:154190735-154420984 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT10 | NM_198321.3 | NP_938080.1 | ||
AB078145.1 | BAC56890.1 | |||
AJ505950.1 | CAD44532.1 | |||
AK023782.1 | BAB14676.1 | |||
AK127135.1 | ||||
BC072450.1 | AAH72450.1 | |||
BC098444.1 | AAH98444.1 |
Target Gene Details
Entrez Gene ID: | 386627 |
Gene Name: | SAP30L antisense RNA 1 (head to head) |
Gene Aliases: |
GALNT10-AS1 |
Location: |
Chr.5:154389769-154445822 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAP30L-AS1 | NR_037897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830520 | Chr.5:154372285 - 154553755 on Build GRCh38 | Loss | GALNT10 SAP30L-AS1 HAND1 SAP30L |
nsv1015362 | Chr.5:154215521 - 154450098 on Build GRCh38 | Gain | MIR1294 GALNT10 SAP30L-AS1 SAP30L |
nsv5080 | Chr.5:154381371 - 154406233 on Build GRCh38 | Insertion | GALNT10 SAP30L-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |