Assay Details
Target Gene Details
Entrez Gene ID: | 9867 |
Gene Name: | praja ring finger ubiquitin ligase 2 |
Gene Aliases: |
Neurodap1, RNF131 |
Location: |
Chr.5:109334709-109409974 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PJA2 | NM_014819.4 | NP_055634.3 | ||
XM_017010099.1 | XP_016865588.1 | |||
AB007898.1 | ||||
AK291759.1 | ||||
BC030826.1 | AAH30826.1 | |||
CR749579.1 | CAH18371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv599338 | Chr.5:109193028 - 109340036 on Build GRCh38 | Gain | PJA2 FER LOC285638 |
esv2763481 | Chr.5:109269689 - 109357995 on Build GRCh38 | Gain | PJA2 LOC285638 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |